Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336936.1 | NP_001323674.1 | 10 - 11 | 1849 | 113 | |
NM_129738.5 | NP_565958.1 | 10 - 11 | 1619 | 113 | ||
GenBank mRNA | AK226882.1 | - | 10 - 11 | 1314 | 113 | |
AY080601.1 | - | 10 - 11 | 1379 | 113 | ||
AY133782.1 | - | 9 - 10 | 1176 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |