Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336936.1 | NP_001323674.1 | 15 - 16 | 2412 | 71 | |
NM_129738.5 | NP_565958.1 | 15 - 16 | 2182 | 71 | ||
GenBank mRNA | AK226882.1 | - | 15 - 16 | 1877 | 71 | |
AY080601.1 | - | 15 - 16 | 1942 | 71 | ||
AY133782.1 | - | 14 - 15 | 1739 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |