Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336936.1 | NP_001323674.1 | 8 - 9 | 1600 | 68 | |
NM_129738.5 | NP_565958.1 | 8 - 9 | 1370 | 68 | ||
GenBank mRNA | AK226882.1 | - | 8 - 9 | 1065 | 68 | |
AY080601.1 | - | 8 - 9 | 1130 | 68 | ||
AY133782.1 | - | 7 - 8 | 927 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |