Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129750.4 | NP_181717.3 | 1 - 2 | 870 | 129 | |
NM_201932.2 | NP_973661.1 | 1 - 2 | 499 | 129 | ||
GenBank mRNA | DQ446617.1 | - | 1 - 2 | 499 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |