Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336961.1 | NP_001324441.1 | 1 - 2 | 561 | 61 | |
NM_129788.3 | NP_181755.1 | 2 - 3 | 598 | 61 | ||
GenBank mRNA | AK227846.1 | - | 2 - 3 | 528 | 61 | |
BT005212.1 | - | 1 - 2 | 387 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |