Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129802.6 | NP_565972.1 | 3 - 4 | 456 | 98 | |
GenBank mRNA | AB125257.1 | - | 1 - 2 | 209 | 98 | |
AF361831.1 | - | 3 - 4 | 368 | 98 | ||
AY078048.1 | - | 1 - 2 | 209 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |