Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129804.4 | NP_565973.1 | 1 - 2 | 618 | 78 | |
GenBank mRNA | AB473851.1 | - | 1 - 2 | 489 | 78 | |
AF345339.1 | - | 1 - 2 | 491 | 78 | ||
AF410334.1 | - | 1 - 2 | 529 | 78 | ||
AF447890.1 | - | 1 - 2 | 514 | 78 | ||
AK230268.1 | - | 1224 | 78 | |||
BT000866.1 | - | 1 - 2 | 303 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |