Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129840.5 | NP_181807.1 | 6 - 7 | 855 | 100 | |
GenBank mRNA | AK227008.1 | - | 6 - 7 | 832 | 100 | |
AY048214.1 | - | 6 - 7 | 831 | 100 | ||
AY124841.1 | - | 6 - 7 | 773 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |