Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129841.4 | NP_181808.1 | 1 | 2105 | 145 | |
GenBank mRNA | BT023464.1 | - | 1 | 1427 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |