Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPMEPCRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129866.5 | NP_181833.1 | 1 - 2 | 1026 | 69 | |
GenBank mRNA | BT023433.1 | - | 1 - 2 | 894 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |