Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT-P4H-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337014.1 | NP_001324186.1 | 3 - 4 | 388 | 75 | |
NM_129869.5 | NP_181836.1 | 3 - 4 | 481 | 75 | ||
GenBank mRNA | AK117688.1 | - | 3 - 4 | 258 | 75 | |
BT006098.1 | - | 3 - 4 | 174 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |