Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129897.4 | NP_181864.1 | 3 - 4 | 613 | 75 | |
GenBank mRNA | BT024899.1 | - | 3 - 4 | 490 | 75 | |
L34413.1 | - | 3 - 4 | 550 | 75 | ||
U31806.1 | - | 3 - 4 | 547 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |