Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFQO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337026.1 | NP_001324650.1 | 9 - 10 | 1258 | 77 | |
NM_129901.4 | NP_181868.1 | 9 - 10 | 1295 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |