Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084577.2 | NP_001078046.1 | 2 - 3 | 782 | 103 | |
NM_001084578.1 | NP_001078047.1 | 2 - 3 | 770 | 103 | ||
NM_001084579.2 | NP_001078048.1 | 2 - 3 | 782 | 103 | ||
NM_001337027.1 | NP_001323584.1 | 2 - 3 | 1933 | 103 | ||
NM_001337028.1 | NP_001323585.1 | 2 - 3 | 772 | 103 | ||
NM_129902.3 | NP_181869.2 | 2 - 3 | 782 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |