Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084577.2 | NP_001078046.1 | 4 - 5 | 1672 | 100 | |
NM_001084578.1 | NP_001078047.1 | 4 - 5 | 1660 | 100 | ||
NM_001084579.2 | NP_001078048.1 | 4 - 5 | 1672 | 100 | ||
NM_001337027.1 | NP_001323584.1 | 4 - 5 | 2823 | 100 | ||
NM_001337028.1 | NP_001323585.1 | 5 - 6 | 1533 | 100 | ||
NM_129902.3 | NP_181869.2 | 5 - 6 | 1543 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |