Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: COS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129967.4 | NP_181933.1 | 5 - 6 | 629 | 66 | |
GenBank mRNA | AF148649.1 | - | 5 - 6 | 538 | 66 | |
AY050913.1 | - | 5 - 6 | 578 | 66 | ||
AY091343.1 | - | 5 - 6 | 538 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |