Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084585.2 | NP_001078054.1 | 10 - 11 | 1104 | 86 | |
NM_129973.3 | NP_181938.1 | 10 - 11 | 1327 | 86 | ||
GenBank mRNA | AF361801.1 | - | 10 - 11 | 1094 | 86 | |
AK226690.1 | - | 10 - 11 | 1097 | 86 | ||
AK227643.1 | - | 10 - 11 | 1094 | 86 | ||
AK316933.1 | - | 10 - 11 | 1098 | 86 | ||
AY050418.2 | - | 10 - 11 | 1095 | 86 | ||
AY113036.1 | - | 10 - 11 | 992 | 86 | ||
D83531.1 | - | 10 - 11 | 1095 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |