Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASHH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129978.4 | NP_566010.1 | 3 - 4 | 563 | 67 | |
GenBank mRNA | AY045886.1 | - | 3 - 4 | 450 | 67 | |
AY091447.1 | - | 3 - 4 | 269 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |