Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345714.1 | NP_001331432.1 | 2 - 3 | 416 | 62 | |
NM_001345715.1 | NP_001331431.1 | 2 - 3 | 416 | 62 | ||
NM_001345716.1 | NP_001331433.1 | 1 - 2 | 513 | 62 | ||
NM_148163.3 | NP_680468.1 | 2 - 3 | 495 | 62 | ||
GenBank mRNA | BT046193.1 | - | 1 - 2 | 302 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |