Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201734.1 | NP_973463.1 | 1 | 1034 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |