Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G09100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_202534.2 | NP_974263.1 | 13 - 14 | 1812 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |