Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G60540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340049.1 | NP_001327136.1 | 2 | 417 | 150 | |
NM_115918.3 | NP_191613.1 | 2 | 382 | 150 | ||
NM_202738.2 | NP_974467.1 | 1 | 811 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |