Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127166.4 | NP_028242.1 | 1 - 2 | 537 | 148 | |
GenBank mRNA | AB252219.1 | - | 1 - 2 | 273 | 148 | |
AY080829.1 | - | 1 - 2 | 274 | 148 | ||
AY088807.1 | - | 1 - 2 | 382 | 148 | ||
AY113976.1 | - | 1 - 2 | 273 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |