Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128872.3 | NP_180871.1 | 1 - 2 | 545 | 127 | |
GenBank mRNA | AF419564.1 | - | 1 - 2 | 325 | 127 | |
AY072422.1 | - | 1 - 2 | 328 | 127 | ||
AY079037.1 | - | 1 - 2 | 187 | 127 | ||
AY086363.1 | - | 1 - 2 | 393 | 127 | ||
AY114706.1 | - | 1 - 2 | 187 | 127 | ||
AY128288.1 | - | 1 - 2 | 325 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |