Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331303.1 | NP_001320442.1 | 1 - 2 | 569 | 149 | |
NM_100058.2 | NP_171680.1 | 2 - 3 | 611 | 149 | ||
GenBank mRNA | AY072410.1 | - | 2 - 3 | 362 | 149 | |
AY088305.1 | - | 2 - 3 | 433 | 149 | ||
BT001250.1 | - | 1 - 2 | 271 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |