Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331338.1 | NP_001321619.1 | 2 - 3 | 493 | 115 | |
NM_100124.5 | NP_171745.3 | 2 - 3 | 493 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |