Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101978.5 | NP_564137.1 | 1 - 2 | 1181 | 156 | |
GenBank mRNA | AY039917.1 | - | 1 - 2 | 1086 | 156 | |
BT001967.1 | - | 1 - 2 | 840 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |