Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117582.4 | NP_193232.1 | 1 - 2 | 273 | 109 | |
NM_179057.1 | NP_849388.1 | 1 - 2 | 208 | 109 | ||
GenBank mRNA | AK316754.1 | - | 1 - 2 | 204 | 109 | |
AY058858.1 | - | 1 - 2 | 203 | 109 | ||
AY065347.1 | - | 1 - 2 | 203 | 109 | ||
AY079036.1 | - | 1 - 2 | 97 | 109 | ||
AY117174.1 | - | 1 - 2 | 97 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |