Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179228.2 | NP_849559.1 | 1 - 2 | 753 | 128 | |
GenBank mRNA | AY150394.1 | - | 1 - 2 | 672 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |