Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtLPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333289.1 | NP_001322048.1 | 1 - 2 | 632 | 117 | |
NM_001333290.1 | NP_001322049.1 | 2 - 3 | 522 | 117 | ||
NM_001333291.1 | NP_001322047.1 | 2 - 3 | 522 | 117 | ||
NM_103699.5 | NP_175237.1 | 1 - 2 | 632 | 117 | ||
NM_179451.1 | NP_849782.1 | 1 - 2 | 274 | 117 | ||
GenBank mRNA | AF228639.3 | - | 1 - 2 | 274 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |