Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084338.2 | NP_001077807.1 | 2 - 3 | 803 | 123 | |
NM_179544.2 | NP_849875.1 | 2 - 3 | 799 | 123 | ||
GenBank mRNA | AY070107.1 | - | 2 - 3 | 771 | 123 | |
AY102107.1 | - | 2 - 3 | 770 | 123 | ||
AY123016.1 | - | 2 - 3 | 732 | 123 | ||
BT000581.1 | - | 2 - 3 | 732 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |