Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336310.1 | NP_001323510.1 | 3 - 4 | 1813 | 179 | |
NM_001336311.1 | NP_001323511.1 | 3 - 4 | 1796 | 179 | ||
NM_001336312.1 | NP_001323509.1 | 3 - 4 | 1746 | 179 | ||
NM_179825.3 | NP_850156.1 | 3 - 4 | 1615 | 179 | ||
GenBank mRNA | AF135189.1 | - | 3 - 4 | 1538 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |