Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G37480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179952.3 | NP_850283.1 | 2 - 3 | 858 | 117 | |
GenBank mRNA | AK176407.1 | - | 2 - 3 | 806 | 117 | |
AK176430.1 | - | 2 - 3 | 806 | 117 | ||
AK176785.1 | - | 2 - 3 | 822 | 117 | ||
AK220984.1 | - | 2 - 3 | 839 | 117 | ||
AK226918.1 | - | 2 - 3 | 841 | 117 | ||
AK229913.1 | - | 2 - 3 | 839 | 117 | ||
AK230011.1 | - | 2 - 3 | 809 | 117 | ||
AK230020.1 | - | 2 - 3 | 839 | 117 | ||
AK230147.1 | - | 2 - 3 | 809 | 117 | ||
AK230323.1 | - | 2 - 3 | 840 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |