Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COR15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202804.1 | NP_001189733.1 | 2 - 3 | 223 | 151 | |
NM_001336986.1 | NP_001323750.1 | 2 - 3 | 350 | 151 | ||
NM_129815.5 | NP_181782.1 | 1 - 2 | 519 | 151 | ||
NM_180040.3 | NP_850371.1 | 2 - 3 | 223 | 151 | ||
GenBank mRNA | AY057640.1 | - | 1 - 2 | 279 | 151 | |
X64138.1 | - | 261 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |