Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180045.4 | NP_850376.2 | 2 - 3 | 208 | 154 | |
GenBank mRNA | AK228536.1 | - | 2 - 3 | 141 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |