Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121688.4 | NP_197184.2 | 1 - 2 | 511 | 172 | |
NM_180501.3 | NP_850832.1 | 1 - 2 | 513 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |