Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180739.2 | NP_851070.1 | 1 - 2 | 1138 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |