Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105299.3 | NP_176802.1 | 10 - 11 | 1413 | 150 | |
GenBank mRNA | AY074378.1 | - | 10 - 11 | 1204 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |