Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106318.4 | NP_177794.1 | 2 - 3 | 881 | 139 | |
GenBank mRNA | AY074874.1 | - | 2 - 3 | 595 | 139 | |
BT020365.1 | - | 2 - 3 | 525 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |