Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103296.4 | NP_174832.1 | 1 - 2 | 114 | 159 | |
GenBank mRNA | AB473866.1 | - | 1 - 2 | 114 | 159 | |
DQ056480.1 | - | 1 - 2 | 101 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |