Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333367.1 | NP_001321457.1 | 3 - 4 | 783 | 170 | |
NM_001333368.1 | NP_001321458.1 | 2 - 3 | 879 | 170 | ||
NM_001333369.1 | NP_001321455.1 | 3 - 4 | 868 | 170 | ||
NM_001333370.1 | NP_001321456.1 | 1 - 2 | 510 | 170 | ||
NM_001333371.1 | NP_001321459.1 | 2 - 3 | 369 | 170 | ||
NM_103806.2 | NP_564541.1 | 3 - 4 | 570 | 170 | ||
NM_179456.3 | NP_849787.1 | 4 - 5 | 727 | 170 | ||
GenBank mRNA | AB085617.1 | - | 3 - 4 | 353 | 170 | |
AF360136.1 | - | 3 - 4 | 570 | 170 | ||
AK317094.1 | - | 4 - 5 | 560 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |