Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104504.3 | NP_176021.1 | 2 - 3 | 576 | 151 | |
GenBank mRNA | AK175677.1 | - | 2 - 3 | 524 | 151 | |
BT010431.1 | - | 2 - 3 | 373 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |