Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP72A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112326.4 | NP_188083.1 | 2 - 3 | 576 | 154 | |
GenBank mRNA | AK228721.1 | - | 2 - 3 | 501 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |