Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203028.2 | NP_001189957.1 | 4 - 5 | 1756 | 154 | |
NM_113282.3 | NP_189020.2 | 4 - 5 | 1729 | 154 | ||
GenBank mRNA | AY862891.1 | - | 4 - 5 | 1724 | 154 | |
AY935711.1 | - | 3 - 4 | 1528 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |