Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113642.4 | NP_189364.1 | 2 - 3 | 951 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |