Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115059.3 | NP_190768.1 | 1 - 2 | 411 | 75 | |
GenBank mRNA | DQ056620.1 | - | 1 - 2 | 350 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |