Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116969.3 | NP_567344.1 | 3 - 4 | 908 | 128 | |
GenBank mRNA | AF412067.1 | - | 3 - 4 | 819 | 128 | |
AY065064.1 | - | 3 - 4 | 852 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |