Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABB1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117820.2 | NP_193449.1 | 4 - 5 | 672 | 143 | |
GenBank mRNA | DQ056652.1 | - | 4 - 5 | 475 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |