Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119665.3 | NP_195225.1 | 1 - 2 | 524 | 164 | |
GenBank mRNA | AY519612.1 | - | 1 - 2 | 264 | 164 | |
BT024907.1 | - | 1 - 2 | 264 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |