Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATB' ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120427.3 | NP_195967.1 | 1 - 2 | 1619 | 150 | |
GenBank mRNA | BT002933.1 | - | 1 - 2 | 1509 | 150 | |
U73526.1 | - | 1 - 2 | 1461 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |